CH4182075119 ChemBridge Corporation

4-(3-methoxyphenoxy)aniline

  • MDL number MFCD02663342
  • Cas Number : 56705-86-3
  • Molecular Weight : 215.2478

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4008767

Safety Information

MSDS https://www.hit2lead.com/sds/4008767_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart