CH4182075143 ChemBridge Corporation

2-(chloromethyl)-4,6-dimethylpyrimidine

  • MDL number MFCD09864267
  • Cas Number : 74502-83-3
  • Molecular Weight : 156.6128

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4009660

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4009660
Availability details for
Quantity
  • Availability
  • Add to Cart