CH4182075195 ChemBridge Corporation

6-methylquinoline-4-carboxylic acid

  • MDL number MFCD00784840
  • Cas Number : 816448-94-9
  • Molecular Weight : 187.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4010352

Safety Information

MSDS https://www.hit2lead.com/sds/4010352_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart