CH4182075208 ChemBridge Corporation

(2-oxopiperidin-1-yl)acetic acid

  • MDL number MFCD00962837
  • Cas Number : 72253-28-2
  • Molecular Weight : 157.1671

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4010683

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4010683
Availability details for
Quantity
  • Availability
  • Add to Cart