CH4182075225 ChemBridge Corporation

5-methyl-1H-pyrazole-3-carbaldehyde

  • MDL number MFCD09055186
  • Cas Number : 3273-44-7
  • Molecular Weight : 110.1139

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4002200

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4002200
Availability details for
Quantity
  • Availability
  • Add to Cart