CH4182075309 ChemBridge Corporation

1-(4-fluorophenyl)propan-1-amine

  • MDL number MFCD07186373
  • Cas Number : 74877-10-4
  • Molecular Weight : 153.1967

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4012664

Safety Information

MSDS https://www.hit2lead.com/sds/4012664_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart