CH4182075326 ChemBridge Corporation

2,2-dimethyl-2,3-dihydro-1-benzofuran-5-carbaldehyde

  • MDL number MFCD08445737
  • Cas Number : 38002-92-5
  • Molecular Weight : 176.2118

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4000012

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4000012
Availability details for
Quantity
  • Availability
  • Add to Cart