CH4182075354 ChemBridge Corporation

4-oxo-1-adamantyl acetate

  • MDL number MFCD01825828
  • Cas Number : 63382-10-5
  • Molecular Weight : 208.2536

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4013897

Safety Information

MSDS https://www.hit2lead.com/sds/4013897_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart