CH4182075407 ChemBridge Corporation

1-(1-propylpiperidin-4-yl)methanamine

  • MDL number MFCD07186440
  • Cas Number : 392691-05-3
  • Molecular Weight : 156.2685

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4014887

Safety Information

MSDS https://www.hit2lead.com/sds/4014887_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart