CH4182075417 ChemBridge Corporation

1-methyl-6-oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid

  • MDL number MFCD06496437
  • Cas Number : 33548-32-2
  • Molecular Weight : 156.1393

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4000132

Safety Information

MSDS https://www.hit2lead.com/sds/4000132_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart