CH4182075418 ChemBridge Corporation

4-(4-fluorophenyl)tetrahydro-2H-pyran-4-carboxylic acid

  • MDL number MFCD08144103
  • Cas Number : 473706-11-5
  • Molecular Weight : 224.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4014951

Safety Information

MSDS https://www.hit2lead.com/sds/4014951_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart