CH4182075454 ChemBridge Corporation

2,5-dimethyl-1H-indole-3-carbaldehyde

  • MDL number MFCD06800613
  • Cas Number : 61364-25-8
  • Molecular Weight : 173.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4002638

Safety Information

MSDS https://www.hit2lead.com/sds/4002638_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart