CH4182075482 ChemBridge Corporation

ethyl cyclohexyl(oxo)acetate

  • MDL number MFCD09055348
  • Cas Number : 13275-31-5
  • Molecular Weight : 184.2322

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4016274

Safety Information

MSDS https://www.hit2lead.com/sds/4016274_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart