CH4182075483 ChemBridge Corporation

3H-imidazo[4,5-b]pyridine-2-thiol

  • MDL number MFCD01088736
  • Cas Number : 29448-81-5
  • Molecular Weight : 151.189

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4016422

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/building-blocks/4016422
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart