CH4182075487 ChemBridge Corporation

1-(1-adamantyl)propan-2-amine

  • MDL number MFCD02632259
  • Cas Number : 39978-69-3
  • Molecular Weight : 193.3284

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4002778

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4002778
Availability details for
Quantity
  • Availability
  • Add to Cart