CH4182075505 ChemBridge Corporation

(2-methyl-1,3-thiazol-4-yl)methanol

  • MDL number MFCD08703637
  • Cas Number : 76632-23-0
  • Molecular Weight : 129.1802

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4016970

Safety Information

MSDS https://www.hit2lead.com/sds/4016970_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart