CH4182075512 ChemBridge Corporation

1-(3-chloro-4-methylphenyl)piperidin-4-one

  • MDL number MFCD09055351
  • Cas Number : 938459-09-7
  • Molecular Weight : 223.6987

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4017221

Safety Information

MSDS https://www.hit2lead.com/sds/4017221_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart