CH4182075527 ChemBridge Corporation

(isoquinolin-5-ylmethyl)methylamine

  • MDL number MFCD08572159
  • Cas Number : 157610-84-9
  • Molecular Weight : 172.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Storage temp. -20
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4002850

Safety Information

MSDS https://www.hit2lead.com/sds/4002850_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart