CH4182075571 ChemBridge Corporation

1-(8-methoxyquinolin-5-yl)-N-methylmethanamine

  • MDL number MFCD08572161
  • Cas Number : 937647-97-7
  • Molecular Weight : 202.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4003027

Safety Information

MSDS https://www.hit2lead.com/sds/4003027_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart