CH4182075585 ChemBridge Corporation

6-(chloromethyl)-2-methylpyrimidin-4-ol

  • MDL number MFCD00129130
  • Cas Number : 35252-96-1
  • Molecular Weight : 158.5856

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4020216

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4020216
Availability details for
Quantity
  • Availability
  • Add to Cart