CH4182075606 ChemBridge Corporation

1-ethyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD02090883
  • Cas Number : 400858-54-0
  • Molecular Weight : 140.1399

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4003135

Safety Information

MSDS https://www.hit2lead.com/sds/4003135_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart