CH4182075613 ChemBridge Corporation

3-ethylmorpholine

  • MDL number MFCD09054774
  • Cas Number : 55265-24-2
  • Molecular Weight : 115.1735

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4021665

Safety Information

MSDS https://www.hit2lead.com/sds/4021665_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart