CH4182075999 ChemBridge Corporation

1-(3-aminophenyl)-5-methyl-2-pyrrolidinone

  • MDL number MFCD10686558
  • Cas Number : 1033693-04-7
  • Molecular Weight : 190.2417

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4027147

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4027147
Availability details for
Quantity
  • Availability
  • Add to Cart