CH4182076026 ChemBridge Corporation

2-(ethylamino)-1-propanol

  • MDL number MFCD11053920
  • Cas Number : 24417-04-7
  • Molecular Weight : 103.1628

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4027506

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4027506
Availability details for
Quantity
  • Availability
  • Add to Cart