CH4182076158 ChemBridge Corporation

4,5-dihydro-1,4-benzoxazepin-3(2H)-one

  • MDL number MFCD01546835
  • Cas Number : 34844-80-9
  • Molecular Weight : 163.1733

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://www.hit2lead.com/building-blocks/4027041

Safety Information

msds https://www.hit2lead.com/building-blocks/4027041
Availability details for
Quantity
  • Availability
  • Add to Cart