CH4182076209 ChemBridge Corporation

methyl 6-formyl-2-pyridinecarboxylate

  • MDL number MFCD07367925
  • Cas Number : 69950-65-8
  • Molecular Weight : 165.1461

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4027144

Safety Information

MSDS https://www.hit2lead.com/sds/4027144_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart