CH4182076217 ChemBridge Corporation

1,3-benzothiazole-2,6-diamine

  • MDL number MFCD00613407
  • Cas Number : 5407-51-2
  • Molecular Weight : 165.2156

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4027157

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4027157
Availability details for
Quantity
  • Availability
  • Add to Cart