CH4182076232 ChemBridge Corporation

3-amino-3-oxopropanoic acid

  • MDL number MFCD00154605
  • Cas Number : 2345-56-4
  • Molecular Weight : 103.0767

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4027194

Safety Information

MSDS https://www.hit2lead.com/sds/4027194_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart