CH4182076305 ChemBridge Corporation

7-amino-1,8-naphthyridin-2(1H)-one

  • MDL number MFCD09743308
  • Cas Number : 1931-44-8
  • Molecular Weight : 161.1607

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4027370

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4027370
Availability details for
Quantity
  • Availability
  • Add to Cart