CH4182076327 ChemBridge Corporation

N-(2-hydroxyethyl)-N-methylglycine

  • MDL number MFCD09709947
  • Cas Number : 26294-19-9
  • Molecular Weight : 133.1457

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4027421

Safety Information

MSDS https://www.hit2lead.com/sds/4027421_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart