CH4182076374 ChemBridge Corporation

1-[4-(3-fluorophenyl)tetrahydro-2H-pyran-4-yl]methanamine

  • MDL number MFCD10686572
  • Cas Number : 943109-41-9
  • Molecular Weight : 209.2599

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://www.hit2lead.com/building-blocks/4027547

Safety Information

msds https://www.hit2lead.com/building-blocks/4027547
Availability details for
Quantity
  • Availability
  • Add to Cart