CH4182076378 ChemBridge Corporation

(2,5-dioxo-1-propyl-4-imidazolidinyl)acetic acid

  • MDL number MFCD07391220
  • Cas Number : 1008961-08-7
  • Molecular Weight : 200.1919

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4027556

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4027556
Availability details for
Quantity
  • Availability
  • Add to Cart