CH4182076454 ChemBridge Corporation

1-methyl-1H-indole-4-carbaldehyde

  • MDL number MFCD08690261
  • Cas Number : 133994-99-7
  • Molecular Weight : 159.1846

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4027737

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4027737
Availability details for
Quantity
  • Availability
  • Add to Cart