CH4182076455 ChemBridge Corporation

(2-amino-1,1-dimethylethyl)dimethylamine

  • MDL number MFCD05201592
  • Cas Number : 76936-44-2
  • Molecular Weight : 116.2046

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4027739

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4027739
Availability details for
Quantity
  • Availability
  • Add to Cart