CH4182076484 ChemBridge Corporation

[5-(4-aminophenyl)-2-furyl]methanol

  • MDL number MFCD01952800
  • Cas Number : 54146-51-9
  • Molecular Weight : 189.2105

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4027829

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4027829
Availability details for
Quantity
  • Availability
  • Add to Cart