CH4182076504 ChemBridge Corporation

1-(4-morpholinyl)acetone

  • MDL number MFCD08690805
  • Cas Number : 6704-35-4
  • Molecular Weight : 143.1836

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4027871

Safety Information

MSDS https://www.hit2lead.com/sds/4027871_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart