CH4182076524 ChemBridge Corporation

1-isopropyl-2,5-dimethyl-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD06380317
  • Cas Number : 18870-77-4
  • Molecular Weight : 165.2322

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4027942

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4027942
Availability details for
Quantity
  • Availability
  • Add to Cart