CH4182076551 ChemBridge Corporation

(3-sec-butoxybenzyl)amine

  • MDL number MFCD09991465
  • Cas Number : 37806-41-0
  • Molecular Weight : 179.2588

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4026828

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4026828
Availability details for
Quantity
  • Availability
  • Add to Cart