CH4182076559 ChemBridge Corporation

3-(cyclopentyloxy)aniline

  • MDL number MFCD09036966
  • Cas Number : 653604-38-7
  • Molecular Weight : 177.2429

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4028025

Safety Information

MSDS https://www.hit2lead.com/sds/4028025_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart