CH4182076588 ChemBridge Corporation

5-propyl-2-furaldehyde

  • MDL number MFCD08144844
  • Cas Number : 14497-27-9
  • Molecular Weight : 138.1638

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4028093

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4028093
Availability details for
Quantity
  • Availability
  • Add to Cart