CH4182076627 ChemBridge Corporation

3-[(methylamino)methyl]phenol

  • MDL number MFCD09733571
  • Cas Number : 123926-62-5
  • Molecular Weight : 137.179

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4028180

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4028180
Availability details for
Quantity
  • Availability
  • Add to Cart