CH4182076648 ChemBridge Corporation

3-methoxy-4-(1-pyrrolidinyl)benzaldehyde

  • MDL number MFCD07403409
  • Cas Number : 116209-27-9
  • Molecular Weight : 205.253

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4028225

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4028225
Availability details for
Quantity
  • Availability
  • Add to Cart