CH4182076707 ChemBridge Corporation

2-(4-morpholinyl)-5-pyrimidinecarbaldehyde

  • MDL number MFCD06589848
  • Cas Number : 842974-69-0
  • Molecular Weight : 193.2025

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4028362

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4028362
Availability details for
Quantity
  • Availability
  • Add to Cart