CH4182076723 ChemBridge Corporation

N-(2-aminoethyl)isonicotinamide

  • MDL number MFCD01332312
  • Cas Number : 17704-88-0
  • Molecular Weight : 165.1924

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4028392

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4028392
Availability details for
Quantity
  • Availability
  • Add to Cart