CH4182076737 ChemBridge Corporation

N-(2-aminoethyl)propanamide

  • MDL number MFCD03308692
  • Cas Number : 925-58-6
  • Molecular Weight : 116.1616

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4028425

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4028425
Availability details for
Quantity
  • Availability
  • Add to Cart