CH4182076793 ChemBridge Corporation

4-amino-2-hydroxy-5-pyrimidinecarboxylic acid

  • MDL number MFCD00047368
  • Cas Number : 3650-93-9
  • Molecular Weight : 155.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4028564

Safety Information

MSDS https://www.hit2lead.com/sds/4028564_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart