CH4182076853 ChemBridge Corporation

1-ethyl-3-piperidinamine

  • MDL number MFCD00044587
  • Cas Number : 6789-94-2
  • Molecular Weight : 128.2153

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4028708

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4028708
Availability details for
Quantity
  • Availability
  • Add to Cart