CH4182076888 ChemBridge Corporation

5-amino-1-cyclohexyl-1H-pyrazol-3-ol

  • MDL number MFCD02855448
  • Cas Number : 436088-86-7
  • Molecular Weight : 181.2349

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4026988

Safety Information

MSDS https://www.hit2lead.com/sds/4026988_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart