CH4182076889 ChemBridge Corporation

tricyclo[3.2.1.0~2,4~]octane-3-carboxylic acid

  • MDL number MFCD08056150
  • Cas Number : 1212345-30-6
  • Molecular Weight : 152.1904

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4028852

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4028852
Availability details for
Quantity
  • Availability
  • Add to Cart