CH4182076988 ChemBridge Corporation

1-[3-(3-fluorophenyl)-5-isoxazolyl]methanamine

  • MDL number MFCD10686612
  • Cas Number : 1033693-12-7
  • Molecular Weight : 192.1897

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4029118

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4029118
Availability details for
Quantity
  • Availability
  • Add to Cart